missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Respiratory Protection, N5500 Elastomeric Halfmask, Class 1
Click to view available options
Size:
Large
Medium
Small
Unit Size:
Each
Pack of 12
Beschreibung
- Lightweight and low-profile ergonomic design encourages high levels of user acceptance
- Unique cradle head band and yoke piece for ease of adjustment
- Contoured revert seal ensures an excellent face seal that will not cause discomfort
- Thermoplastic elastomer material avoids latex allergens that are common with economy masks
- Single unit, four-point harness with neck strap is easy to use
- Harness's ergonomic design and improved suspension system fits perfectly on top of the head and offers a high level of comfort
- Superior exhalation valve allows more breath to be exhaled makes workers feel cooler and more comfortable
- Reduced dead air space means less re-breathed exhaled air
- Variable thickness in sealing area conforms to nose bridge
- Filter cartridges not included, available for order separately
Bestellinformation
Filter cartridges not included, available for order separately
Spezifikation
Spezifikation
| Cartridge/Filter | Filter Cartridges Sold Separately |
| Cartridge Type | Dual Cartridge: Class 1 |
| Certifications/Compliance | EC Category PPE: 3, EC Certificate: 251, EC Attestation: CE Ceert 251-5500 Respir-Dec 2007, EN 140, Declaration of conformity INSPEC N65550031 |
| Color | Gray |
| Material | Thermoplastic Elastomer |
| Size | Medium |
| For Use With (Application) | Chemical, Laboratory, and Petrochemical Industries |
| Includes | 4-Point Head Harness |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur