missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Respiratory Protection, N5500 Elastomeric Halfmask, Class 1

Product Code. 15836845
Change view
Click to view available options
Size:
Large
Medium
Small
Unit Size:
Each
Pack of 12
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15836845 Large Each
15816845 Small Each
15858244 Medium Pack of 12
15826845 Medium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15836845 Supplier Honeywell™ Supplier No. N65550033
  • ONLINE EXCLUSIVE PRICE
    31.61 €
    27.76 € / Pack of 1
    Save 3.85 €
    12% Off
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-Performance, Dual-Cartridge (Class 1) Half-Mask Respirator

  • Lightweight and low-profile ergonomic design encourages high levels of user acceptance
  • Unique cradle head band and yoke piece for ease of adjustment
  • Contoured revert seal ensures an excellent face seal that will not cause discomfort
  • Thermoplastic elastomer material avoids latex allergens that are common with economy masks
  • Single unit, four-point harness with neck strap is easy to use
  • Harness's ergonomic design and improved suspension system fits perfectly on top of the head and offers a high level of comfort
  • Superior exhalation valve allows more breath to be exhaled makes workers feel cooler and more comfortable
  • Reduced dead air space means less re-breathed exhaled air
  • Variable thickness in sealing area conforms to nose bridge
  • Filter cartridges not included, available for order separately

Order Info

Filter cartridges not included, available for order separately

TRUSTED_SUSTAINABILITY

Specifications

Cartridge/Filter Filter Cartridges Sold Separately
Cartridge Type Dual Cartridge: Class 1
Certifications/Compliance EC Category PPE: 3, EC Certificate: 251, EC Attestation: CE Ceert 251-5500Respir-Dec 2007, EN 140, Declaration of conformity INSPEC N65550031
Color Gray
Material Thermoplastic Elastomer
Size Large
For Use With (Application) Chemical, Laboratory, and Petrochemical Industries
Includes 4-Point Head Harness
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.