Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
Product Code. 15816635
Change view
Click to view available options
Protects against organic vapors and acid gas
Product Code.
15816635 Protects against organic vapors and acid gas
1 options
Product Code. 15816635 Supplier Honeywell™ Supplier No. N06575003L
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The A1B1E1 cartridge is part of the wide choice of cartridges offered to protect against many hazardous gases, vapors in many applications. The range provides high quality and a cost-effective protection.
  • Protects against organic vapors and acid gas
  • Excellent filtration efficiency over extensive period of time
  • Lower profile and lightweight for optimal comfort
  • Low profile does not obstruct the field of vision
  • Threaded connector is easy to attach
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Class 1 plastic cartridge
Certifications/Compliance EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259
For Use With (Application) Protects against organic vapors and acid gas
Quantity 6 Pair/Box, 8 Boxes/Cs.
Weight (Metric) 103.9 g