Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
Product Code. 15806845
Change view
Click to view available options
Includes threaded connector
Product Code.
15806845 Includes threaded connector
1 options
Product Code. 15806845 Supplier Honeywell™ Supplier No. N06575008
  • ONLINE EXCLUSIVE PRICE
    8.03 €
    7.68 € / Pack of 1
    Save 0.35 €
    4% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The P3 cartridge is part of the wide range of high-quality and cost-effective cartridges offered to protect against many hazardous gases, vapors and/or particulates in many applications.
  • Protects against particulates
  • Excellent filtration efficiency over extensive period of time
  • Lower profile and lightweight for optimal comfort
  • Low profile does not obstruct the field of vision
  • Threaded connector is easy to attach
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 0252, EC Attestation: 0252, EN 143, Declaration of conformity INSPEC N06575008
Material ABS - fiberglass paper
For Use With (Application) Agriculture, automotive, aviation, construction, chemical, foundry, general industry, glass, green spaces, industrial cleaning, iron and steel, laboratory, maintenance, medical and pharmaceutical, paper, ship building, textile, utilities, water treatment, welding, wood industries
Includes Threaded connector