Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
Product Code. 15846635
Change view
Click to view available options
Protects against organic vapor and particulates
Product Code.
15846635 Protects against organic vapor and particulates
1 options
Product Code. 15846635 Supplier Honeywell™ Supplier No. N06575081L
  • ONLINE EXCLUSIVE PRICE
    27.96 €
    22.70 € / 1 pairs
    Save 5.26 €
    18% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The A1P3 cartridge is part of the wide range of cartridges offered to protect against many hazardous gases, vapors and/or particulates in many applications. The range provides high quality and a cost-effective protection.
  • Protects against organic vapor and particulates
  • Excellent filtration efficiency over extensive period of time
  • Lower profile and lightweight for optimal comfort
  • Low profile does not obstruct the field of vision
  • Threaded connector is easy to attach
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Class 1 plastic cartridge
Certifications/Compliance EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259
For Use With (Application) Protects against organic vapor and particulates
Quantity 6 Pair/Box, 8 Boxes/Cs.
Weight (Metric) 92.5 g