Learn More
Prestige Medical™ Classic Portable Autoclave Media
Click to view available options
Capacity (Metric):
9 and 12 L
Unit Size:
Each
Description
The availability of a wide range of model sizes, features and internal furniture options make the Prestige Medical Classic a popular choice where a cost effective, portable autoclave is needed. This 9 liter Classic Media autoclave sterilises at 121°C and has a cycle time of approximately 70 minutes. Fully automatic, simply press the button to start. Lightweight and portable, ideal for laboratory use - the appliance for science.
- High quality construction
- Fully automatic electronic time and temperature control
- Steam release valve automatically opens to release excess pressure
- Automatic fault warning light and cut-out if unit boils dry or sterilisation is interrupted
- Dual interlock handles prevent the lid being opened before the pressure and temperature inside have fallen to safe levels
- Complete with instrument basket
- Models 2015 onwards available with optional printer for easy data recording
- CE marked
- Comprehensive one year warranty (UK)
- Interlock safety mechanism, eliminates the risk of super-heated vessels exploding
- Fully automatic, 'Set and forget' - load your flask into the basket, set the programme with a one touch button, relax and forget until the cycle has ended
- Reliable, special in-built software ensures exact temperatures are reached for the correct duration
- 9L and 12L models
- Fits neatly onto the smallest of workbenches For the sterilisation of media, liquids and contaminated waste.
Specifications
Specifications
| Capacity (Metric) | 9 and 12 L |