missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 18.2 Recombinant Human Monoclonal Antibody (9C2)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA543155
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.
Specifications
| Claudin 18.2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P56856 | |
| CLDN18 | |
| Recombinant Human Claudin-18.2 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry | |
| 9C2 | |
| Unconjugated | |
| CLDN18 | |
| SFTA5; SFTPJ | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 51208 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction