missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Claudin 5 Monoclonal Antibody (4C3C2)

Product Code. 10777223
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10777223 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10777223 Supplier Invitrogen™ Supplier No. 352500
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Claudin 5 Monoclonal Antibody for ELISA, IHC (P), Western Blot

This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4C3C2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CLDN5
Gene Accession No. O00501, O54942, Q9JKD6
Gene Alias AI854493; AWAL; BEC1; Brain endothelial cell clone 1 protein; claudin 5; claudin 5 (transmembrane protein deleted in velocardiofacial syndrome); claudin-5; claudin-5 protein; Cldn5; CPETRL1; lung-specific membrane protein; MBEC1; TMDVCF; TMVCF; Transmembrane protein deleted in VCFS; transmembrane protein deleted in velocardiofacial syndrome; Unknown (protein for MGC:138054)
Gene Symbols CLDN5
Host Species Mouse
Immunogen Synthetic peptide derived from the mouse Claudin-5 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12741, 65131, 7122
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.