missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 572.00 €
Specifications
| Antigen | CLCA4 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18475041
|
Novus Biologicals
NBP1-86688-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18248136
|
Novus Biologicals
NBP1-86688 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLCA4 Polyclonal specifically detects CLCA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CLCA4 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| caCC-2, CaCC2CaCC, Calcium-activated chloride channel family member 4, Calcium-activated chloride channel protein 2, calcium-activated chloride channel regulator 4, chloride channel accessory 4, chloride channel regulator 4, chloride channel, calcium activated, family member 4, hCaCC-2, hCLCA4, MGC142247, MGC142249 | |
| CLCA4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 22802 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:WVGMVHFDSTATIVNKLIQIKSSDERNTLMAGLPTYPLGGTSICSGIKYAFQVIGELHSQLDGSEVLLLTDGEDNTASSCIDEV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title