Learn More
Greiner Bio-One μClear™ Bottom 96-well Polystyrene Microplates
Product Code. 10030262
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 40
Product Code. Color Unit Size
10030262 Black Pack of 40
1 options
Product Code. 10030262 Supplier Greiner Bio-One Supplier No. 655096
  • ONLINE EXCLUSIVE PRICE
    507.00 €
    269.95 € / Pack of 40
    Save 237.05 €
    46% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Minimises the risk of sample carryover and cross-contamination. Greiner Bio-One μClear™ Bottom 96-Well Polystyrene Microplates is free of detectable DNase, RNase, human DNA.
  • Non-Pyrogenic
  • Material: Polystyrene
  • Free of detectable DNase, RNase, human DNA
  • Traced way back to production through defined LOT number system
  • More suitable for non polar proteins and peptides
  • Medium binding
Certifications
DIN ISO 9001
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type Medium Binding
Color Black
Product Type Assay Microplate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Medium Binding
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Chimney