missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates
BIOPHARMA

Product Code. 10098860
Change view
Click to view available options
Surface Treatment:
MaxiSorp
PolySorp
Certified:
No
Yes
Unit Size:
Pack of 60
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Surface Treatment Certified Unit Size
10098860 MaxiSorp No Pack of 60
10737931 PolySorp No Pack of 60
10777621 MaxiSorp Yes Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.
Product Code. 10098860 Supplier Thermo Scientific™ Supplier No. 430341
  • ONLINE EXCLUSIVE PRICE
    181.03 €
    149.00 € / Pack of 60
    Save 32.03 €
    17% Off
View all promotions
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Perform quantitative and qualitative solid phase immunoassays with these nonsterile 96-well plates, available in a variety of binding surfaces.

Perform quantitative and qualitative solid phase immunoassays with Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates, which include Nunc™, Immulon™ and Microtiter™ plates. Choose from a variety of binding surfaces for reliable and reproducible results in colormetric assays.

96-Well Solid Plates

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces

  • PolySorp for adsorption of hydrophobic molecules
  • MaxiSorp is hydrophilic and ideal for antibody sandwich assays

Ordering Information:

Please note that availability may vary by country. 446612 includes certificate.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Hydrophilic
Certified No
Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 60 Cases
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 250 μL
Sterility Non-sterile
Surface Treatment MaxiSorp
Well Shape Round, C Bottom
No. per Case 60
No. per Pack 5
Plate Blocking None
Quantity 60/Cs.
Target Molecule Antibodies (Glycoproteins)
Well Design C96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.