missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ cleavage stimulation factor Protein

Product Code. p-200039672 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.5mg
100µg
This item is not returnable. View return policy

Product Code. 18433591

Brand: Novus Biologicals™ NBP1988710.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-431 of Human cleavage stimulation factor The Recombinant Human cleavage stimulation factor Protein is derived from E. coli. The Recombinant Human cleavage stimulation factor Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. 20 mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 0.1 M NaCl, 2 M urea
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Human
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name cleavage stimulation factor
Conjugate Unconjugated
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.