missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CLEC14A Antibody (Scripps Korea patent anti-CLEC14A) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
213.00 € - 920.00 €
Specifications
| Antigen | CLEC14A |
|---|---|
| Clone | Scripps Korea patent anti-CLEC14A |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30046571
|
Novus Biologicals™
NBP3-28399-100ug |
100 μg |
338.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046675
|
Novus Biologicals™
NBP3-28399-1mg |
1 mg |
920.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045441
|
Novus Biologicals™
NBP3-28399-50ug |
50 μg |
213.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLEC14A Monoclonal antibody specifically detects CLEC14A in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| CLEC14A | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Human | |
| Q86T13 | |
| 161198 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
| Scripps Korea patent anti-CLEC14A | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C14orf27, CEG1, chromosome 14 open reading frame 27, ClECT and EGF-like domain containing protein, C-type lectin domain family 14 member A, C-type lectin domain family 14, member A, EGFR5, EGFR-5, Epidermal growth factor receptor 5 | |
| CLEC14A | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title