missing translation for 'onlineSavingsMsg'
Learn More

Clethodim Sulfone, TRC

Product Code. 17960995
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960995 100 mg 100mg
17904915 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17960995 Supplier Toronto Research Chemicals Supplier No. TRCC573260100MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111031-17-5
Molecular Formula C17 H26 Cl N O5 S
Molecular Weight (g/mol) 391.91
Synonym 2-Cyclohexen-1-one, 2-[1-[[(3-chloro-2-propenyl)oxy]imino]propyl]-5-[2-(ethylsulfonyl)propyl]-3-hydroxy- (9CI), 2-[1-[[(3-Chloro-2-propen-1-yl)oxy]imino]propyl]-5-[2-(ethylsulfonyl)propyl]-3-hydroxy-2-cyclohexen-1-one, Clethodim sulfone
IUPAC Name 2-[(E)-N-[(E)-3-chloroprop-2-enoxy]-C-ethylcarbonimidoyl]-5-(2-ethylsulfonylpropyl)-3-hydroxycyclohex-2-en-1-one
SMILES CC\C(=N/OC\C=C\Cl)\C1=C(O)CC(CC(C)S(=O)(=O)CC)CC1=O

Specifications

Analyte or Component Names Clethodim Sulfone
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H26ClNO5S/c1-4-14(19-24-8-6-7-18)17-15(20)10-13(11-16(17)21)9-12(3)25(22,23)5-2/h6-7,12-13,20H,4-5,8-11H2,1-3H3/b7-6+,19-14+
Solution Type Neat
Industry Type Forensics
Quantity 100 mg
Formula Weight 391.122
Chemical Name or Material Clethodim Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.