missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Click-iT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit
GREENER_CHOICE

Product Code. 11380952
Change view
Click to view available options
Quantity:
100 assays
50 assays
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380952 100 assays Each
11300962 50 assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380952 Supplier Molecular Probes™ Supplier No. C10420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Click-iT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit

The Click-iT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit provides a simplified, more robust assay for analyzing DNA replication in proliferating cells as compared to traditional BrdU methods. Newly synthesized DNA is analyzed using the 488 nm laser of the flow cytometer.

  • Accurate—Superior results compared to BrdU assays
  • Fast—results in as little as 90 minutes
  • Economical—More assays per kit

    View selection guide for all Click-iT™ EdU and Click-iT™ Plus EdU assays for flow cytometry.

    An Advanced Method Giving You Results Superior to BrdU
    The most accurate method of proliferation analysis is direct measurement of DNA synthesis. Originally, this was performed through incorporation of radioactive nucleosides, i.e. 3H-thymidine. This method was replaced by antibody-based detection of the nucleoside analog bromodeoxyuridine (BrdU). The Click-iT™ EdU Flow Cytometry Assay Kits are novel alternatives to the BrdU assay. EdU (5-ethynyl-2´-deoxyuridine) is a thymidine analog which is incorporated into DNA during active DNA synthesis. Detection is based on click chemistry; a copper catalyzed covalent reaction between an azide and an alkyne. In this application, the alkyne is found in the ethynyl moiety of EdU, while the azide is coupled to Alexa Fluor ™ 488, Alexa Fluor™ 647, or Pacific BlueTM dyes. Standard flow cytometry methods are used for determining the percentage of S-phase cells in the population (Fig 1).

    Mild Conditions Allow Use with Cell Cycle Dyes and Antibodies
    The advantages of Click-iT™ EdU labeling are readily evident while performing the assay (Fig 2). The small size of the dye azide allows for efficient detection of the incorporated EdU using mild conditions, while standard aldehyde-based fixation and detergent permeabilization are sufficient for the Click-iT™ detection reagent to gain access to the DNA. This is in contrast to BrdU assays that require DNA denaturation (using HCl, heat, or digestion with DNase) to expose the BrdU so that it may be detected with an anti-BrdU antibody. Sample processing for the BrdU assay can result in signal alteration of the cell cycle distribution as well as destruction of antigen recognition sites when using the HCl method. In contrast, the easy-to-use EdU cell proliferation kit is compatible with cell cycle dyes. This EdU assay kit can also be multiplexed with antibodies against surface and intracellular markers.

    Quick and Simple Protocol
    The Click-iT™ EdU protocol is based on the aldehyde fixation and detergent permeabilization steps for immunohistochemical antibody labeling, but EdU is compatible with other
    fixation/permeabilization agents including saponin and methanol. In just five steps you’ll be ready to analyze your cell proliferation data:

  • Treat cells with EdUM
  • Fix and permeabilize cells
  • Detect S-phase cells with Click-iT™ detection cocktail for 30 min
  • Wash once
  • Analyze

    Get Accurate Results Economically
    By increasing the number of assays per kit, the Click-IT™ EdU Alexa Fluor™ 488 Flow Cytometry Assay Kit is less expensive than the traditional BrdU assays making them ideal for large experiments.

    For research use only. Not intended for any human or animal therapeutic of diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Contains EdU (5-ethynyl-2' -deoxyuridine), Alexa Fluor™ 488 azide, anhydrous dimethylsulfoxide (DMSO), Click-iT™ fixative, Click-iT™ saponin-based permeabilization and wash buffer, copper (II) sulfate,, and Click-iT™ EdU buffer additive.
  • Store at 2°C to 8°C
  • Dessicate and protect from light.
    Detection Method Fluorescence
    For Use With (Equipment) Flow Cytometer
    Product Type Flow Cytometry Assay Kit
    Dye Type Alexa Fluor™ 488
    Emission 488
    Format Tube(s)
    Product Line Alexa Fluor, Click-iT
    Quantity 100 assays
    Shipping Condition Room Temperature

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.