missing translation for 'onlineSavingsMsg'
Learn More

Climbazole, MedChemExpress

Product Code. 16442256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442256 500 mg 500mg
16438646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442256 Supplier Medchem Express Supplier No. HYB1151
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Climbazole (BAY-e 6975) is a potent antifungal agent. Climbazole also is a potent inducer of rat hepatic cytochrome P450.

Climbazole (BAY-e 6975) is a potent antifungal agent. Climbazole also is a potent inducer of rat hepatic cytochrome P450.

Specifications

Chemical Name or Material Climbazole
Physical Form Solid
CAS 38083-17-9
Color Off-White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C15H17ClN2O2
For Use With (Application) COVID-19-immunoregulation
Synonym BAY-e 6975
Solubility Information DMSO : ≥ 47 mg/mL (160.54 mM)
SMILES CC(C)(C)C(C(OC1=CC=C(Cl)C=C1)N2C=CN=C2)=O
Molecular Weight (g/mol) 292.76
Formula Weight 292.76
Percent Purity 98.9%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.