missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Clindamycin hydrochloride, 97%

Product Code. 15595106 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15595106 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15595106 Supplier Thermo Scientific Chemicals Supplier No. 443510100
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Clindamycin hydrochloride, CAS # 21462-39-5, is a lincosamide antibiotic. | CAS: 21462-39-5 | C18H34Cl2N2O5S | 461.439 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Clindamycin is a lincosamide antibiotic derived from lincomycin that has antibiotic activity against aerobic gram-positive cocci, and parasitic protozoa
  • Clindamycin reversibly binds to the 50s bacterial ribosomal subunit, thus inhibiting protein synthesis

Applications

  • Clindamycin has been used to enhance bacterial colonization in research mice during the study of commensal gut microbiota
  • Clindamycin has been used to study biofilm-mediated antibiotic resistance in an in vitro model of G. vaginalis
  • Clindamycin was used as a selection agent to isolate, enumerate, and identify Lactobacillus spp. from probiotic dairy products
TRUSTED_SUSTAINABILITY

Specifications

Type Clindamycin Hydrochloride
CAS 21462-39-5
Melting Point 156.0°C to 159.0°C
Infrared Spectrum Authentic
Water 6% max.
Quantity 10 mg
Molecular Formula C18H34Cl2N2O5S
Solubility Information Solubility in water: ca 50g/L. Other solubilities: soluble in pyridine, ethanol, dmf
InChI Key AUODDLQVRAJAJM-PIPVPTKYSA-N
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl
IUPAC Name (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 461.439
PubChem CID 134129519
Formula Weight 461.45
Percent Purity 97%
Chemical Name or Material Clindamycin hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Clindamycin hydrochloride
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.