missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin hydrochloride monohydrate

Product Code. 15474009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474009 1 g 1g
15484009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474009 Supplier Thermo Scientific Alfa Aesar Supplier No. J61409.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Clindamycin hydrochloride, CAS # 21462-39-5, is the salt of clindamycin, a derivative of the broad-spectrum bacteriostatic antibiotic lincomycin. | CAS: 58207-19-5 | C18H34Cl2N2O5S | 461.439 g/mol

Clindamycin hydrochloride monohydrate is an antibacterial agent that is a semisynthetic analog of lincomycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Clindamycin is a lincosamide antibiotic that disrupts protein synthesis by interfering with the aminoacyl-tRNA ribosome complex

• Clindamycin decreases sebaceous gland activity and blocks the neuromuscular junction

Application

• Clindamycin is commonly used in laboratory applications to inhibit the growth anaerobic microbes, but has some effectiveness against aerobic microbes as well

• It can be used in the laboratory study of cell culture for resistance testing, or in antibiotic allergic reaction studies

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58207-19-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD07793327
InChI Key AUODDLQVRAJAJM-NFOLQJLNSA-N
PubChem CID 131632848
IUPAC Name (2S,4R)-N-[2-chloro-1-[(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Type Clindamycin Hydrochloride Monohydrate
Physical Form Powder
Color White
Quantity 1 g
Beilstein 4070786
Merck Index 14,2356
Solubility Information Soluble in water
Formula Weight 479.46 (461.44 Anhydrous)
Chemical Name or Material Clindamycin hydrochloride monohydrate
compliance-icons
Product Identifier
  • Clindamycin hydrochloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Clindamycin hydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.