missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP170 Antibody (CL4851) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61144JF669
This item is not returnable.
View return policy
Description
CLIP170 Monoclonal antibody specifically detects CLIP170 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| CLIP170 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CAP-GLY domain containing linker protein 1, CLIP, CLIP170, CLIP-170Reed-Sternberg intermediate filament-associated protein, CYLN1MGC131604, cytoplasmic linker 1, Cytoplasmic linker protein 1, Cytoplasmic linker protein 170 alpha-2, Reed-Steinberg cell-expressed intermediate filament-associated protein, Restin, restin (Reed-Steinberg cell-expressed intermediate filament-associated protein), RSNCAP-Gly domain-containing linker protein 1 | |
| This antibody was developed using a recombinant protein derived from P30622, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Dendritic Cell Markers | |
| 6249 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL4851 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction