missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLK1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17190244 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17190244 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17190244 Supplier Invitrogen™ Supplier No. PA5112388

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CLK1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

CLK1 is a dual specificity kinase that phosphorylates PTP-1B and members of the SR family of splicing factors. It is thought to be involved in neuronal differentiation and mRNA splicing. Therefore, the encoded protein may play an indirect role in governing splice site selection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLK1
Gene Accession No. P49759
Gene Alias CDC like kinase 1; CDC28/CDC2-like kinase; CDC-like kinase 1; CLK; CLK/STY; CLK1; Dual specificity protein kinase CLK1; Protein kinase CLK1; protein kinase STY; protein tyrosine kinase STY; Sty
Gene Symbols CLK1
Host Species Rabbit
Immunogen Recombinant Human Dual specificity protein kinase CLK1 protein (1-130AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1195
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.