missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate, 95+%

Product Code. 15493119
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493119 5 g 5g
15483119 1 g 1g
15403129 25 g 25g
15413129 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15493119 Supplier Thermo Scientific Alfa Aesar Supplier No. J60342.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 637-07-0 | C12H15ClO3 | 242.70 g/mol

It is a lipid-lowering agent used for controlling the high cholesterol and triacylglyceride level in the blood. It increases lipoprotein lipase activity to promote the conversion of VLDL to LDL, and hence reduce the level of VLDL. It can increase the level of HDL as well. Clofibrate is a ligand agonist for PPARα (peroxisome proliferator-activated receptor α), or an inducer of cytochromes P450’s CYP4A1 and CYP4A3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble to 100 mM in DMSO.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat, bases. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-07-0
Molecular Formula C12H15ClO3
Molecular Weight (g/mol) 242.70
MDL Number MFCD00000615
InChI Key KNHUKKLJHYUCFP-UHFFFAOYSA-N
Synonym clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
PubChem CID 2796
ChEBI CHEBI:3750
IUPAC Name ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1

Specifications

Density 1.14
Boiling Point 154°C to 156°C
Flash Point 113°C (235°F)
Refractive Index 1.5
Quantity 5 g
UN Number UN3082
Beilstein 1913459
Merck Index 14,2377
Solubility Information Soluble to 100 mM in DMSO.
Formula Weight 242.7
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material Clofibrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.