missing translation for 'onlineSavingsMsg'
Learn More

Clofibric Acid, TRC

Product Code. 17983085
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983085 100 mg 100mg
17914055 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17983085 Supplier Toronto Research Chemicals Supplier No. TRCC586930100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-09-7
Molecular Formula C10 H11 Cl O3
Molecular Weight (g/mol) 214.645
Synonym Propionic acid, 2-(p-chlorophenoxy)-2-methyl- (6CI,7CI,8CI), 2-(4-Chlorophenoxy)-2-methylpropanoic acid, (p-Chlorophenoxy)isobutyric acid, 2-(4-Chlorophenoxy)-2-methylpropanic acid, 2-(4-Chlorophenoxy)-2-methylpropionic acid, 2-(p-Chlorophenoxy)-2-methylpropanoic acid, 2-(p-Chlorophenoxy)-2-methylpropionic acid, 2-(p-Chlorophenoxy)isobutyric acid, 2-[(4-Chlorophenyl)oxy]-2-methylpropionic acid, 4-Chlorophenoxyisobutyric acid, Arteriohom, CPIB, CPIBA, Chlorfibrinic acid, Chlorofibrinic acid, Chlorophibrinic acid, Clofibrate free acid, Clofibric acid, Clofibrilic acid, Clofibrin, Clofibrinic acid, NSC 1149, PCIB, PCPIB, Regadrin, Regulipid, α-(4-Chlorophenoxy)-α-methylpropionic acid, α-(4-Chlorophenoxy)isobutyric acid, α-(p-Chlorophenoxy)isobutyric acid
IUPAC Name 2-(4-chlorophenoxy)-2-methylpropanoic acid
SMILES CC(C)(Oc1ccc(Cl)cc1)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H11ClO3/c1-10(2,9(12)13)14-8-5-3-7(11)4-6-8/h3-6H,1-2H3,(H,12,13)
Formula Weight 214.04
Chemical Name or Material 2-(4-Chlorophenoxy)-2-methylpropionic acid (Clofibric acid)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.