missing translation for 'onlineSavingsMsg'
Learn More

Clomipramine hydrochloride, MedChemExpress

Product Code. 16488316
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488316 100 mg 100mg
16432146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488316 Supplier Medchem Express Supplier No. HYB0457
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clomipramine (Chlorimipramine) hydrochloride is a potent 5-HT reuptake blocker with the IC50 value of 1.5 nM. Clomipramine hydrochloride is a tricyclic antidepressant that can be used for the research of depression and obsessive compulsive disorder (OCD).

Clomipramine (Chlorimipramine) hydrochloride is a potent 5-HT reuptake blocker with the IC50 value of 1.5 nM. Clomipramine hydrochloride is a tricyclic antidepressant that can be used for the research of depression and obsessive compulsive disorder (OCD).

Specifications

Chemical Name or Material Clomipramine hydrochloride
Physical Form Solid
CAS 17321-77-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H24Cl2N2
For Use With (Application) Neuroscience-Neuromodulation
Synonym Chlorimipramine hydrochlorideG-34586 hydrochlorideNSC-169865 hydrochloride
Solubility Information DMSO : 50 mg/mL (142.32 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (142.32 mM)
SMILES CN(C)CCCN1C2=CC(Cl)=CC=C2CCC3=C1C=CC=C3.Cl
Molecular Weight (g/mol) 351.31
Formula Weight 351.31
Percent Purity 99.49%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.