missing translation for 'onlineSavingsMsg'
Learn More

Clopidol, MedChemExpress

Product Code. 16412256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412256 500 mg 500mg
16408606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16412256 Supplier Medchem Express Supplier No. HYB1088
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clopidol (WR-61112) is an anticoccidial agent which is used as feed additive to control coccidiosis in chickens. Clopidol inhibits the sporulation of Eimeria tenella oocysts.

Clopidol (WR-61112) is an anticoccidial agent which is used as feed additive to control coccidiosis in chickens. Clopidol inhibits the sporulation of Eimeria tenella oocysts.

Specifications

Chemical Name or Material Clopidol
Physical Form Solid
CAS 2971-90-6
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C7H7Cl2NO
For Use With (Application) COVID-19-immunoregulation
Synonym WR-61112
Solubility Information 0.1 M NaOH : 26 mg/mL (135.39 mM; ultrasonic and adjust pH to 3 with NaOH) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OC1=C(Cl)C(C)=NC(C)=C1Cl
Molecular Weight (g/mol) 192.04
Formula Weight 192.04
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.