missing translation for 'onlineSavingsMsg'
Learn More

Closantel, MedChemExpress

Product Code. 16491886
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491886 500 mg 500mg
16467386 100 mg 100mg
16421276 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491886 Supplier Medchem Express Supplier No. HY17596
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Closantel is a halogenated salicylanilide with a potent anti-parasitic activity. Closantel is a potent and highly specific Onchocerca volvulus chitinase (OvCHT1) inhibitor with an IC50 of 1.6 μM and a Ki of 468 nM. Closantel inhibits the O. volvulus L3 to L4 molt of developing.

Closantel

Specifications

Chemical Name or Material Closantel
Physical Form Solid
CAS 57808-65-8
Color Off-White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C22H14Cl2I2N2O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 49 mg/mL (73.90 mM) ∣H2O : 1 mg/mL (1.51 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(NC1=CC(Cl)=C(C(C2=CC=C(Cl)C=C2)C#N)C=C1C)C3=CC(I)=CC(I)=C3O
Molecular Weight (g/mol) 663.07
Formula Weight 663.07
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.