missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clostridium Difficile Selective Agar

Product Code. 12947138
Click to view available options
Description:
Clostridium Difficile Selective Agar
Quantity:
10/Pk.
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12947138

Brand: Thermo Scientific™ PB0218A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Isolate, identify and differentiate Clostridium difficile using this selective medium which utilizes D-cycloserine and cefoxitin to inhibit non-target organisms.

Isolate, identify and differentiate Clostridium difficile using Thermo Scientific™ Clostridium Difficile Selective Agar, a selective medium with two key components to inhibit non-target organisms.

Selective agents D-cycloserine (500μg/mL) and cefoxitin (16μg/mL) used in the formulation inhibit growth of the majority of Enterobacteriaceae, as well as Streptococcus fecalis, staphylococci, Gram-negative non-sporing anaerobic bacilli, and Clostridium species (except Clostridium difficile), which may be found in large numbers in fecal samples.

Clostridium difficile
is a toxin-producing, sporeforming anaerobic gram-positive rod, which causes gastrointestinal infections ranging in severity from asymptomatic colonization to severe diarrhea, pseudomembranous colitus (PMC), toxic megacolon, colonic perforaton, and death. A reduction of the normal microbial flora in the colon, usually caused by antibiotic therapy, allows overgrowth of C. difficile. Most pathogenic strains of C. difficile produce two toxins, toxin A (enterotoxin) and toxin B (cytotoxin), which are the main virulence factors for the organism. An emerging strain of C. difficile contains an additional binary toxin (CDT) believed to be a marker of increased virulence. This strain is missing a gene that regulates production of toxin A and B and has acquired high-level resistance to fluoroquinolone antibiotics.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Ruby red, opaque
Content And Storage 2°C to 10°C
Description Clostridium Difficile Selective Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.