missing translation for 'onlineSavingsMsg'
Learn More

Clotrimazole, MedChemExpress

Product Code. 16411119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411119 Supplier Medchem Express Supplier No. HY10882
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clotrimazole is an imidazole derivative, an antifungal compound and is a CYP (cytochrome P450) inhibitor. Clotrimazole has antibacterial activity.

Clotrimazole is an imidazole derivative, an antifungal compound and is a CYP (cytochrome P450) inhibitor. Clotrimazole has antibacterial activity.

Specifications

Chemical Name or Material Clotrimazole
Physical Form Solid
CAS 23593-75-1
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C22H17ClN2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (144.99 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES ClC1=CC=CC=C1C(N2C=CN=C2)(C3=CC=CC=C3)C4=CC=CC=C4
Molecular Weight (g/mol) 344.84
Formula Weight 344.84
Percent Purity 99.91%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.