missing translation for 'onlineSavingsMsg'
Learn More

Clotrimazole

Product Code. 15496049
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496049 25 g 25g
15486049 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496049 Supplier Thermo Scientific Alfa Aesar Supplier No. J63895.14
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specific inhibitor of calcium-activated potassium channels | CAS: 23593-75-1 | C22H17ClN2 | 344.842 g/mol

Clotrimazole is an imidazole derivative and an antifungal compound and a CYP (cytochrome P450) inhibitor. Clotrimazole has been shown to block the intermediate-conductance, IK1 channels (Ca2+ activated K+ channels), in cells such as erythrocytes. In vitro studies of various yeast strains have demonstrated susceptibility to clotrimazole. Clotrimazole is an activator of MB67 and an inhibitor of CYP3A4 and CYP51A1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Clotrimazole is an imidazole derivative and an antifungal compound and a CYP (cytochrome P450) inhibitor. Clotrimazole has been shown to block the intermediate-conductance, IK1 channels (Ca2+ activated K+ channels), in cells such as erythrocytes. In vitro studies of various yeast strains have demonstrated susceptibility to clotrimazole. Clotrimazole is an activator of MB67 and an inhibitor of CYP3A4 and CYP51A1.

Solubility
Soluble in chloroform, DMSO, DMF and alcohols

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23593-75-1
Molecular Formula C22H17ClN2
Molecular Weight (g/mol) 344.842
MDL Number MFCD00057220
InChI Key VNFPBHJOKIVQEB-UHFFFAOYSA-N
Synonym 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole; 1-(o-Chlorotrityl)imidazole
PubChem CID 2812
ChEBI CHEBI:3764
IUPAC Name 1-[(2-chlorophenyl)-diphenylmethyl]imidazole
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4

Specifications

Type Clotrimazole
Physical Form Powder
Melting Point 141°C to 149°C
Color White
Quantity 25 g
Merck Index 14,2417
Solubility Information Soluble in chloroform, DMSO, DMF and alcohols
Formula Weight 344.84
Chemical Name or Material Clotrimazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.