missing translation for 'onlineSavingsMsg'
Learn More

Clotrimazole, TRC

Product Code. 30134731
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134731 1 g 1g
30132657 100 mg 100mg
30005556 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134731 Supplier Toronto Research Chemicals Supplier No. TRCC5874001G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23593-75-1
Molecular Formula C22 H17 Cl N2
Molecular Weight (g/mol) 344.84
Synonym Imidazole, 1-(o-chloro-α,α-diphenylbenzyl)- (8CI), 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole, 1-(o-Chlorophenyldiphenylmethyl)imidazole, 1-(o-Chlorotrityl)imidazole, Agisten, BAY 5097, BAY 5907, BAY-B 5097, Canesten, Canifug, Clotrimazol, Clotrimazole, Desamix F, Diphenyl(2-chlorophenyl)(1-imidazolyl)methane, Empecid, Femcare, Gyne-Lotrimin, Gyne-Lotrimin 7, Locasten, Lotrimin, Lotrimin AF Cream, Lotrimin AF Solution, Lotrimin Jock-Itch Cream, Lotrimin Jock-Itch Lotion, Monobaycuten, Mycelex, Mycelex 7, Mycelex G, Mycelex OTC, Mycelex Troche, Mycofug, Mycosporin, NSC 257473, Pedisafe, Plimycol, Rimazole, Tibatin, Trimysten, Veltrim
IUPAC Name 1-[(2-chlorophenyl)-diphenylmethyl]imidazole
SMILES Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4ccnc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H17ClN2/c23-21-14-8-7-13-20(21)22(25-16-15-24-17-25,18-9-3-1-4-10-18)19-11-5-2-6-12-19/h1-17H
Formula Weight 344.108
Percent Purity >95%
Chemical Name or Material Clotrimazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.