missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cloud Point Standard, Paragon Scientific™
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Description
- Cloud Point Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Cloud Point according to IP 219. Alternative test methods are ASTM D2500 and ISO 3015.
- Certified according to ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
Specifications
| Chemical Name or Material | Diesel, UKAS ISO 17034 Certified (-7.7 °C) |
| Certifications/Compliance | IP 219 |
| Quantity | 250 mL |
Safety and Handling
Product Identifier
- Certified Reference Material Cloud Point Standard,
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
- MIXTURE LIST-Contains : diesel
Nominal values quoted
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction