missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cloxacillin sodium salt monohydrate

Product Code. 15356256 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Tamaño de la unidad
15356256 1 g 1g
15565136 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15356256 Proveedor Thermo Scientific Chemicals N.º de proveedor 455300010
ChemsChrom Mega Deals
Use promo code 27993
Ver todas las promociones
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Cloxacillin sodium salt monohydrate, CAS # 7081-44-9, is a compound that targets gram-positive bacteria and shows antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cloxacillin sodium salt monohydrate is a chlorinated derivative of oxacillin that shows β-lactamase-resistant antibiotic activity

• This compound inhibits the last stage of bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs), which results in cell lysis, that is mediated by bacterial cell wall autolytic enzymes

Applications

• Cloxacillin sodium salt monohydrate targets gram-positive bacteria, and is effective against staphylococci that produce β-lactamase

• It acts in vitro against both penicillin gram-resistant and gram-sensitive staphylococci, certain streptococci, and pneumococci

• This compound is commonly used in vivo and in vitro studies as AmpC β-lactamase inhibitor in combination with β-lactam antibiotics

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7081-44-9
Molecular Formula C19H19ClN3NaO6S
Molecular Weight (g/mol) 475.876
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
PubChem CID 23675743
ChEBI CHEBI:34978
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]

Especificaciones

Type Cloxacillin Sodium Salt Monohydrate
Infrared Spectrum Authentic
Water 3.0 to 5.0%
Quantity 1 g
Formula Weight 475.88
Percent Purity ≥94% (HPLC)
Chemical Name or Material Cloxacillin sodium salt monohydrate
compliance-icons compliance-icons
Nombre del producto
  • Cloxacillin sodium salt monohydrate
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.