missing translation for 'onlineSavingsMsg'
Learn More

Cloxacillin Sodium Salt, MP Biomedicals™

Product Code. 11472072 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472072 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11472072 Supplier MP Biomedicals Supplier No. 0215070225
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits the last stage of bacterial cell wall synthesis by binding to certain penicillin-binding proteins (PBPs). Cloxacillin Sodium Salt, MP Biomedicals is a semi-synthetic, β-lactamase-resistant antibiotic used to treat infections. It is a chlorin

Cloxacillin binds to certain penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes. It may interfere with autolysin inhibitors. Cloxacillin has many antibiotic uses:

  • To treat infections caused by penicillin G-resistant staphylococci
  • In studies of the use of topical antibacterial agents for burn and crush wounds
  • As an AmpC β-lactamase inhibitor in combination with β-lactam antibiotics

Specifications

Physical Form Powder
CAS 7081-44-9
pH 6.0 to 7.5 (literature)
Color White to Off-White
Quantity 25 g
Molecular Formula C19H19ClN3NaO6S
Synonym 5-Methyl-3-(o-chlorophenyl)-4-isoxazolyl] pencillin, cloxacillin sodium
Solubility Information Freely soluble in water (200mg + 4 mL, clear colorless to faint yellow solution) and methanol; soluble in alcohol, ethylene gycol, ethyl, and pyridene.
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
Molecular Weight (g/mol) 475.876
PubChem CID 23675743
ChEBI CHEBI:34978
Formula Weight 457.86
Percent Purity 100.8%
Grade Not Available
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cloxacillin Sodium Salt
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Cloxacillin Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.