Learn More
CLP Contract Required Detection Limit (CRDL) Solution 1: Pb @ 6; Be, Cd, Se @ 10; Ag, As, Cr, Tl @ 20; Mn @ 30; Zn @ 40; Cu @ 50; Ni @ 80; Co, V @ 100; Sb @ 120 μg/mL in 5% HNO3, tr. Tartaric Acid, VHG™
Product Code. 30003139
Change view
Click to view available options
Unit Size:
100mL
Product Code. Unit Size
30003139 100mL
1 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003139 Supplier VHG Labs Supplier No. VHGCRDL100
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
CLP Contract Required Detection Limit (CRDL) Solution 1: Pb @ 6; Be, Cd, Se @ 10; Ag, As, Cr, Tl @ 20; Mn @ 30; Zn @ 40; Cu @ 50; Ni @ 80; Co, V @ 100; Sb @ 120 μg/mL in 5% HNO3, tr. Tartaric Acid
TRUSTED_SUSTAINABILITY
Specifications
Solvent or Matrix Dilute HNO3
Quantity 100 mL