missing translation for 'onlineSavingsMsg'
Learn More

CLP Instrument Calibration Standard 1: Ag @ 200; Be @ 400; Cd @ 500; Ba, Co, Cu, Fe, Mn, Ni, Pb, Tl, Zn @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17978669
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17978669 500 mL 500mL
17989139 100 mL 100mL
18060430 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17978669 Supplier VHG Labs Supplier No. VHGICL1500
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

CLP Instrument Calibration Standard 1: Ag @ 200; Be @ 400; Cd @ 500; Ba, Co, Cu, Fe, Mn, Ni, Pb, Tl, Zn @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver/Beryllium/Cadmium/Barium/Cobalt/Copper/Iron/Lead/Manganese/Nickel/Thallium/Zinc
CAS 7440-22-4, 7440-41-7, 7440-43-9, 7440-39-3, 7440-48-4, 7440-50-8
Methods ICP
Analyte or Component Names Silver/Beryllium/Cadmium/Barium/Cobalt/Copper/Iron/Lead/Manganese/Nickel/Thallium/Zinc
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Silver 200 μg/mL/Beryllium 400 μg/mL/Cadmium 500 μg/mL/Barium 1000 μg/mL/Cobalt 1000 μg/mL/Copper 1000 μg/mL/Iron 1000 μg/mL/Lead 1000 μg/mL/Manganese 1000 μg/mL/Nickel 1000 μg/mL/Thallium 1000 μg/mL/Zinc 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.