missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLRN3 Polyclonal Antibody

Product Code. 13200279
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13200279 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13200279 Supplier Invitrogen™ Supplier No. PA526137
  • ONLINE EXCLUSIVE PRICE
    578.00 €
    410.38 € / 400µL
    Save 167.62 €
    29% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CLRN3 Polyclonal Antibody for Flow, Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CLRN3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CLRN3
Gene Accession No. Q8NCR9
Gene Alias clarin 3; clarin-3; CLRN3; TMEM12; transmembrane protein 12; USH3AL1; Usher syndrome type-3A-like protein 1
Gene Symbols CLRN3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 196-222 amino acids from the C-terminal region of human CLRN3.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119467
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.