missing translation for 'onlineSavingsMsg'
Learn More

Clusterin/APOJ Antibody (SAIC-43B-8) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody

267.00 € - 624.00 €

Specifications

Antigen Clusterin/APOJ
Clone SAIC-43B-8
Concentration 1 mg/mL
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18690045
View Documents
Novus Biologicals
NBP3-20109-0.025mg
0.025 mg
267.00 €
0.02mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18664834
View Documents
Novus Biologicals
NBP3-20109-0.2mg
0.2 mg
624.00 €
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Clusterin/APOJ Monoclonal antibody specifically detects Clusterin/APOJ in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
Specifications

Specifications

Clusterin/APOJ
1 mg/mL
Monoclonal
Purified
RUO
Human
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2
Peptide 'ASSIIDELFQDR' derived from Clusterin conjugated to KLH. Clusterin isoform 1 functions as extracellular chaperone that prevents aggregation of nonnative proteins
Primary
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
SAIC-43B-8
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Mass Spectrometry
Unconjugated
Rabbit
Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Dendritic Cell Markers, Hypoxia, Immunology, Innate Immunity
PBS
1191
IgG κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.