missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMTM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84458
This item is not returnable.
View return policy
Description
CMTM1 Polyclonal specifically detects CMTM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CMTM1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| chemokine-like factor super family 1, chemokine-like factor superfamily 1, Chemokine-like factor superfamily member 1, chemokine-like factor-like protein CKLFH1, CKLFH, CKLFH1, CKLFH1a, CKLF-like MARVEL transmembrane domain containing 1, CKLF-like MARVEL transmembrane domain-containing protein 1, CKLFSF1, MGC71870 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CMTM1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TKMRTNLKRFLGVEVERKLSPAKDAY | |
| 0.1 mL | |
| Cytokine Research | |
| 113540 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction