missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CMTM6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16628004
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16628004 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16628004 Supplier Invitrogen™ Supplier No. PA5101525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CKLF6.

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. Han W., Genomics 81:609-617(2003). Ota T., Nat. Genet. 36:40-45(2004). The MGC Project Team; Genome Res. 14:2121-2127(2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CMTM6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CMTM6
Gene Accession No. Q9CZ69, Q9NX76
Gene Alias 2810051A14Rik; AA536733; chemokine-like factor super family 6; chemokine-like factor superfamily 6; chemokine-like factor superfamily member 6; CKLF like MARVEL transmembrane domain containing 6; CKLF-like MARVEL transmembrane domain containing 6; CKLF-like MARVEL transmembrane domain-containing protein 6; Cklfsf6; Cmtm6; Da2-17; LRRGT00102; PRO2219
Gene Symbols CMTM6
Host Species Rabbit
Immunogen A synthesized peptide derived from human CMTM6(Accession Q9NX76), corresponding to amino acid residues A136-A183.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54918, 67213
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.