missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 593.00 €
Specifications
| Antigen | CNOT1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18478521
|
Novus Biologicals
NBP2-32045-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18100564
|
Novus Biologicals
NBP2-32045 |
0.1 mL |
593.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNOT1 Polyclonal specifically detects CNOT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CNOT1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| AD-005, adrenal gland protein AD-005, CCR4-associated factor 1, CCR4-NOT transcription complex subunit 1, CCR4-NOT transcription complex, subunit 1, CDC39DKFZp686E0722, hNOT1, KIAA1007DKFZp686O168, Negative regulator of transcription subunit 1 homolog, NOT1 (negative regulator of transcription 1, yeast) homolog, NOT1FLJ90644, NOT1HFLJ36492 | |
| CNOT1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| A5YKK6 | |
| 23019 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TIETLMRINAHSRGNAPEGLPQLMEVVRSNYEAMIDRAHGGPNFMMHSGISQASEYDDPPGLREKAEYLLREWVNLYHSAAAGRDSTKAFSAFVG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title