missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CNP Monoclonal Antibody (347608)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561265
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
CNP (2',3'-Cyclic Nucleotide 3' Phosphodiesterase) is a Protein Coding gene. Diseases associated with CNP include Leukodystrophy, Hypomyelinating, 20 and Myopia 2, Autosomal Dominant. Among its related pathways are MECP2 and associated Rett syndrome and Glial cell differentiation. Gene Ontology (GO) annotations related to this gene include RNA binding and cyclic nucleotide binding.
Specifications
| CNP | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P09543 | |
| CNP | |
| E. coli-derived recombinant human CNP, Lys24-Cys126. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 347608 | |
| Unconjugated | |
| CNP | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; CNP1; CNP2; CNP-22; CNP-29; CNP-53; CNPase; C-type natriuretic peptide; C-type natriuretic peptide/factor; HLD20; lbab; natriuretic peptide C; natriuretic peptide precursor C; natriuretic peptide precursor type C; natriuretic peptide type C; NPPC; snake venom metalloprotease inhibitor 02D01-like; SVSP15 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 1267 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction