missing translation for 'onlineSavingsMsg'
Learn More

CNQX, MedChemExpress

Product Code. 16498887
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498887 5 mg 5mg
16493456 25 mg 25mg
16492417 50 mg 50mg
16486626 100 mg 100mg
16406308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498887 Supplier Medchem Express Supplier No. HY15066
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CNQX (FG9065) is a potent and competitive AMPA/kainate receptor antagonist with IC50s of 0.3 μM and 1.5 μM, respectively. CNQX is a competitive non-NMDA receptor antagonist. CNQX blocks the expression of fear-potentiated startle in rats.

CNQX (FG9065) is a potent and competitive AMPA/kainate receptor antagonist with IC50s of 0.3 μM and 1.5 μM, respectively. CNQX is a competitive non-NMDA receptor antagonist. CNQX blocks the expression of fear-potentiated startle in rats.

Specifications

Chemical Name or Material CNQX
Physical Form Solid
CAS 115066-14-3
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C9H4N4O4
For Use With (Application) Neuroscience-Neurodegeneration
Synonym FG9065
Solubility Information DMSO : 20 mg/mL (86.15 mM; Need ultrasonic)
SMILES O=C1C(NC2=CC([N+]([O-])=O)=C(C#N)C=C2N1)=O
Molecular Weight (g/mol) 232.15
Formula Weight 232.15
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.