missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTFR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92414-0.1ml
This item is not returnable.
View return policy
Description
CNTFR Polyclonal antibody specifically detects CNTFR in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CNTFR | |
| Polyclonal | |
| Western Blot 1:400-1:2000 | |
| ciliary neurotrophic factor receptor, ciliary neurotrophic factor receptor alpha, ciliary neurotrophic factor receptor subunit alpha, CNTF receptor subunit alpha, CNTFR alpha, CNTFR-alpha, MGC1774 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-342 of human CNTFR (NP_671693.1). FHRDSWHLRHQVLLHVGLPPREPVLSCRSNTYPKGFYCSWHLPTPTYIPNTFNVTVLHGSKIMVCEKDPALKNRCHIRYMHLFSTIKYKVSISVSNALGHNATAITFDEFTIVKPDPPENVVARPVPSNPRRLEVTWQTPSTWPDPESFPLKFFLRYRPLILDQWQHVELSDGTAHTITDAYAGKEYIIQVAAKDNEIGTWSDWSVAAHATPWTEEPRHLTTEAQAAETTTSTTSSLAPPPTTKICDPGELGS | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 1271 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction