Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
2330.00 € - 3340.00 €
Product Code | Brand | Description | Type | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Type | For Use With (Equipment) | Price | Quantity & Availability | |||
11334222
|
Thermo Scientific™
CO4567 |
CO2 Backup System | Factory-Installed | TSX Series, Forma FDE, HERAfreeze HDE, Revco RDE and TDE Series |
2640.00 €
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
11326401
|
Thermo Scientific™
LN4567 |
LN2 Backup System | Factory-Installed | TSX Series, Forma FDE, HERAfreeze HDE, Revco RDE and TDE Series (400 box capacity and larger) |
2500.00 €
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||
16245041
|
Thermo Scientific™
1960445 |
CO2 Backup System | CO2 Backup System | Forma™ 900, 7000 and 8600 Series, HFU B and HFC Series, Revco™ CxF Series, TSC Series |
2330.00 €
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||
16349693
|
Thermo Scientific™
1960446 |
CO2 Backup System | Field-Installed | TSE Series, Forma™ 7000 Series, HFU B and HFC Series, Revco™ CxF Series, TSC Series |
2330.00 €
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
11386381
|
Thermo Scientific™
FCO300D |
CO2 Backup System | Field-Installed | 208-230V/60Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 |
2500.00 €
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
11396381
|
Thermo Scientific™
FCO300V |
CO2 Backup System | CO2 Backup System | 230V/50Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 |
2500.00 €
Each |
In Stock
Log in to see stock. |
|||
11306391
|
Thermo Scientific™
FCO300W |
CO2 Backup System | Field-Installed | 230V/50Hz models (UK Plug): Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 |
2930.00 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
11316391
|
Thermo Scientific™
FCO4567 |
CO2 Backup System | Field-Installed | TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE and TLE Series (400 box and above) |
2480.00 €
Each |
In Stock
Log in to see stock. |
|||
11376391
|
Thermo Scientific™
FLN300D |
LN2 Backup System | Standalone | 208-230V/60Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 |
2500.00 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||
11386391
|
Thermo Scientific™
FLN300V |
LN2 Backup System | LN2 Backup System | 230V/50Hz models: Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 |
2500.00 €
Each |
In Stock
Log in to see stock. |
|||
11396391
|
Thermo Scientific™
FLN300W |
LN2 Backup System | Standalone | 230V/50Hz models (UK Plug): Forma FDE30086, Revco RDE30086, HERAfreeze HDE30086, TDE30086 |
2930.00 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
11306401
|
Thermo Scientific™
FLN4567 |
LN2 Backup System | Field-Installed | Revco™ UxF, HERAfreeze™ HFU-T, Thermo Scientific™ TSU Series |
2500.00 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
15738557
|
Thermo Scientific™
FLN4567LE |
LN2 Backup System: maintains temperature down to -80°C with liquid nitrogen | Field-installed | Thermo Scientific TSX Series |
2500.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
15728557
|
Thermo Scientific™
FCO4567LE |
CO2 Backup System: maintains temperature down to -67°C with CO2 | Field-installed | Thermo Scientific TSX Series |
2500.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
16317141
|
Thermo Scientific™
FCO456LEV |
CO2 Backup System: maintains temperature down to -67°C with CO2 | CO2 Backup System | Forma FDE, Revco RDE, HERAfreeze HDE, and Thermo Scientific TDE Series (400 box capacity and larger) |
2970.00 €
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16367141
|
Thermo Scientific™
FLN456LEV |
LN2 Backup System: maintains temperature down to -80°C with LN2 | LN2 Backup System | Forma FDE, Revco RDE, HERAfreeze HDE, and Thermo Scientific TDE Series (400 box capacity and larger) |
2970.00 €
Each |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17838010
|
Thermo Scientific™
332353G01 |
LN2 Backup System | Factory Installed | TDE Series Chest Freezers |
3340.00 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
17848010
|
Thermo Scientific™
332353G02 |
CO2 Backup System | Factory Installed | TDE Series Chest Freezers |
3110.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
17878010
|
Thermo Scientific™
332353G05 |
LN2 Backup System | Factory Installed | TDE Series Chest Freezers |
3340.00 €
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||
17888010
|
Thermo Scientific™
332353G06 |
CO2 Backup System | Factory Installed | TDE Series Chest Freezers |
3110.00 €
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:- LN2 backup system maintains temperature down to -80°C with liquid nitrogen
- CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.
Specifications
-80°C |