missing translation for 'onlineSavingsMsg'
Learn More

Coatomer protein complex, subunit zeta 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

280.00 € - 589.00 €

Specifications

Antigen Coatomer protein complex, subunit zeta 2
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18482662
View Documents
Novus Biologicals
NBP2-30435-25ul
25ul
280.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18183603
View Documents
Novus Biologicals
NBP2-30435
0.1 mL
589.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Coatomer protein complex, subunit zeta 2 Polyclonal specifically detects Coatomer protein complex, subunit zeta 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Coatomer protein complex, subunit zeta 2
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
coatomer protein complex, subunit zeta 2, coatomer subunit zeta-2, MGC23008, nonclathrin coat protein zeta-COP, zeta-2 coat protein, Zeta-2 COP, Zeta-2-coat protein, zeta2-COP
COPZ2
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Polyclonal
Rabbit
Human
Q9P299
51226
This antibody was developed against a recombinant protein corresponding to amino acids: DPQQVIQKVNFRADDGGLTEQSVAQV
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.