missing translation for 'onlineSavingsMsg'
Learn More

Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits

Product Code. 11313968
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313968 2 kg 2kg
11303968 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313968 Supplier Thermo Scientific Alfa Aesar Supplier No. 040387.A3
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61789-51-3 | C20H34CoO4 | 397.42 g/mol

Cobalt naphthenate is widely used in polyester resins and paint driers. It is used as a curing accelerator, cross linker catalyst for unsaturated polyester resins. It is also used in the production of adhesives, varnishes and waterproofing agents for textiles. Further, it is used as corrosion inhibitors, lubricants, and fuel additives. It plays an important role as oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61789-51-3
Molecular Formula C20H34CoO4
Molecular Weight (g/mol) 397.42
MDL Number MFCD02094763
InChI Key XSWKLHINRKWMTD-UHFFFAOYNA-L
Synonym cobalt naphthenate, cobalt ii 2-naphthoate
PubChem CID 119077305
IUPAC Name cobalt;naphthalene-2-carboxylic acid
SMILES [Co++].CCC1CCC(CCC([O-])=O)C1.CCC1CCC(CCC([O-])=O)C1

Specifications

Flash Point 40°C (104°F)
Quantity 2 kg
UN Number UN1993
Solubility Information Insoluble in water.
Concentration or Composition (by Analyte or Components) Co 6%, approximately 53% in min.eral spirits
Physical Form Liquid
Chemical Name or Material Cobalt naphthenate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.