missing translation for 'onlineSavingsMsg'
Learn More

Cobicistat, MedChemExpress

Product Code. 16460357
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460357 50 mg 50mg
16436717 5 mg 5mg
16414008 10 mg 10mg
16435006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16460357 Supplier Medchem Express Supplier No. HY10493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cobicistat is a potent and selective inhibitor of cytochrome P450 3A (CYP3A) enzymes with IC50s of 30-285 nM. Cobicistat is a pharmacokinetic enhancer which increases the overall absorption of several HIV medications.

Cobicistat is a potent and selective inhibitor of cytochrome P450 3A (CYP3A) enzymes with IC50s of 30-285 nM. Cobicistat is a pharmacokinetic enhancer which increases the overall absorption of several HIV medications.

Specifications

Chemical Name or Material Cobicistat
Physical Form Solid
CAS 1004316-88-4
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C40H53N7O5S2
For Use With (Application) COVID-19-anti-virus
Synonym GS-9350
Solubility Information DMSO : 250 mg/mL (322.16 mM; Need ultrasonic)
SMILES CN(C(N[C@@H](CCN1CCOCC1)C(N[C@@H](CC2=CC=CC=C2)CC[C@@H](NC(OCC3=CN=CS3)=O)CC4=CC=CC=C4)=O)=O)CC5=CSC(C(C)C)=N5
Molecular Weight (g/mol) 776.02
Formula Weight 776.02
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.