Learn More
Palintest™ COD Reagent Test Tubes
These COD reagent test tubes, with a range of 0 to 2000mg/L, are ideal for use in most laboratory and scientific environments where measuring COD.
90.37 € - 188.68 €
Product Code | Brand | Range | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Description | Price | Quantity & Availability | ||||
11918437
|
Palintest™
PL454 |
0 to 2000mg/L | Mercury Free |
90.37 €
25 tests |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11946473
|
Palintest™
PL464 |
0 to 2000mg/L | With Mercury |
188.68 €
Pack of 24 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11928437
|
Palintest™
PL452 |
0 to 400mg/L | — |
96.04 €
Pack of 25 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Ready to use reagent test tubes
- COD reagent test tubes
- Quantity: 1
- Number of Tests: 25
- Range: 0 to 2000mg/L
- Type: COD reagent test tubes
Specifications
25 | |
1 Each | |
COD Reagent Test Tube |