missing translation for 'onlineSavingsMsg'
Learn More

COD Standard, 1000mg/L, Hach Lange™

Product Code. 10178134
Change view
Click to view available options
Quantity:
200 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10178134 200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10178134 Supplier Hach Supplier No. 2253929
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hach Lange COD Standard, 1000mg/L, Quantity: 200mL, Concentration: 1000mg/L, Traceability to NIST: NIST

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1000 mg/L
Quantity 200 mL
compliance-icons
Product Identifier
  • COD Standard Solution, 1000 mg/L
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydrogen phthalate, copper sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.