missing translation for 'onlineSavingsMsg'
Learn More

COD Standard Solution, 400mg/L, WTW™

Product Code. 15277388
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15277388 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15277388 Supplier WTW Supplier No. 252451
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For few parameters such as nitrite, which are subject to a quick conversion, freshly prepared control standards of stock solutions versus the multi-parameter standard checks are mandatory. Stock solutions are also more cost-effective than the already confectioned CombiChecks™. With stock solutions, the correct dilution must be self-calculated and conducted. The use of standards is supported via AQA functions in the photoLab™ photometers.
  • Free chlorine
  • Bound chlorine
  • Formaldehyde
  • Hydrazine
  • Hydrogen peroxide
  • Hydrogen sulfide
  • Phenol
  • Silicon
  • Sulfide
  • Sulfite
  • Anionic surfactants
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Concentration 100 mg/L
Color Green/White
Quantity 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.