missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98910-50ul
This item is not returnable.
View return policy
Description
Cofilin 2 Polyclonal antibody specifically detects Cofilin 2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Cofilin 2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunoprecipitation 1-4 μL/mg of lysate, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| cofilin 2 (muscle), Cofilin, muscle isoform, cofilin-2, NEM7 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Cofilin 2. | |
| 50 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 1073 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction